Total RNA Kits for the quick and easy isolation of high quality total RNA from animal or human cells and tissue.
- Suitable for the simultaneous processing of multiple samples
- No need for organic extractions or time-consuming alcohol precipitations
- Immediate inactivation of exogenous and endogenous RNases
- Favoured binding of RNA molecules >200 bases
- Efficient separation of enzyme inhibitors and contaminants
- Final elution in RNase-free water
- Isolated total RNA directly suitable for downstream applications
*Safety Line columns with snap-on lids. Classic Line columns without lid.
Delivery information: Each kit contains PerfectBind RNA columns (Safety or Classic Line*), collection tubes, DNA Removing columns, RNA Lysis Buffer T, RNA Wash Buffer I and II, and RNase-free water. Advanced protocol for isolation of miRNA included in the manual.