Cycle-Pure kits for quick and reliable purification of up to 30 µg DNA from PCR reactions and other reaction mixtures. DNA purified with peqGOLD Cycle-Pure kits can be directly used for downstream applications such as T/A ligations, sequencing reactions, restriction digestions or labelling reactions.
- Extraction of fragments between 50 bp and 40 kbp
- Efficient removal of enzymes, nucleotides, oligonucleotides, salts and other contaminants
- Easy protocol and short hands-on times of less than 14 minutes
- Suitable for the simultaneous processing of multiple samples
- No need for organic extractions or time-consuming alcohol precipitations
- No shearing of DNA or carry-over of silica material
- Final elution with Elution Buffer or water
*Safety Line columns with snap-on lids, Classic Line columns without lids.
Delivery information: Each kit contains PerfectBind DNA columns (Safety or Classic Line*) and collection tubes, CP Buffer and CG Wash Buffer, and Elution Buffer.