Cycle-Pure Kits, peqGOLD
PEQLEMR502200EA 119 EUR
PEQLEMR502200 PEQL12-6493-01 PEQL12-6493-02 PEQL12-6493-00 PEQLEMR502050 PEQLEMR502005
Cycle-Pure Kits, peqGOLD
Cycle-Pure kits for quick and reliable purification of up to 30 µg DNA from PCR reactions and other reaction mixtures. DNA purified with peqGOLD Cycle-Pure kits can be directly used for downstream applications such as T/A ligations, sequencing reactions, restriction digestions or labelling reactions.
  • Extraction of fragments between 50 bp and 40 kbp
  • Efficient removal of enzymes, nucleotides, oligonucleotides, salts and other contaminants
  • Easy protocol and short hands-on times of less than 14 minutes
  • Suitable for the simultaneous processing of multiple samples
  • No need for organic extractions or time-consuming alcohol precipitations
  • No shearing of DNA or carry-over of silica material
  • Final elution with Elution Buffer or water
*Safety Line columns with snap-on lids, Classic Line columns without lids.
Delivery information: Each kit contains PerfectBind DNA columns (Safety or Classic Line*) and collection tubes, CP Buffer and CG Wash Buffer, and Elution Buffer.