Blood DNA mini kit plus, peqGOLD
PEQL12-3484-02EA 374 EUR
PEQL12-3484-02 PEQLEMR524200 PEQL12-3484-01 PEQLEMR524005 PEQLEMR524050
Blood DNA mini kit plus, peqGOLD
Blood DNA Mini Kit Plus offers a safe, quick and efficient method for extracting pure and high quality DNA from fresh, frozen and anticoagulated treated blood.
  • Extraction of DNA up to 60 kbp
  • Suitable for human or animal blood
  • Suitable for the simultaneous processing of multiple samples
  • No need for time-consuming alcohol precipitations
  • Complete enzymatic degradation of cellular RNA
  • Efficient separation of enzyme inhibitors and contaminants
  • Final elution of DNA in low salt buffer or water
*Safety Line columns with snap-on lids, Classic Line columns without lids.
Delivery information: Kits contain PerfectBind DNA columns (Safety or Classic Line*) and collection tubes, Lysis Buffer BL, Binding Solution BL and TE Buffer, Proteinase K, DNA Wash Buffer BL I and II, and Elution Buffer.