Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Ag-AK, Latex
SearchPresentationType-HORIZONTAL
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
Supplier:  THERMO FISHER DIAGNOSTICS GMBH
Description:   Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
Supplier:  THERMO FISHER DIAGNOSTICS GMBH
Description:   SALMONELLA H POLYVALENT PHASES 1 AND 2 1 * 2 ml
Supplier:  THERMO FISHER DIAGNOSTICS GMBH
Description:   The technique of reversed passive latex agglutination (RPLA) enables soluble antigen such as bacterial toxins to be detected in an agglutination assay...
Catalog Number: (710-0213)
Supplier:  MAST DIAGNOSTICA
Description:   For the identification of Staphylococci, which synthezise the Clumping factor and/or Protein A.
UOM:  1 * 120 Tests
Supplier:  THERMO FISHER DIAGNOSTICS GMBH
Description:   Wellcolex® Rapid Latex Agglutination test kits are designed for rapid and accurate identification of enteric bacteria, such as Salmonella, S...
Supplier:  THERMO FISHER DIAGNOSTICS GMBH
Description:   Directly detect antigens present in human cerebrospinal fluid (CSF), serum, urine, or blood cultures using Thermo Scientific™ Wellcogen™ Bacterial Ant...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6  of 6