Anti-MUC13 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA322118EA 564 EUR
ORIGTA322118
Anti-MUC13 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000-5000; Immunochemistry: 1:25-100

Type: Primary
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR