Coverslipper with tape for slides, CM1260, VWR®
Supplier: VWR, part of Avantor
SERMCM-1260
Coverslipper with tape for slides, CM1260, VWR®
This high-capacity tape coverslipper can cover up to 1260 slides per hour, thanks to a flexible mechanical process.
  • Multilingual touch screen with parameter settings
  • Automatic drying of slides with integrated suction process
  • Adjustable tape length (45 to 70 mm) and dosage of the xylene
  • User-friendly interface with clear overview