You Searched For: TLC Plates
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: Baker-flex® pre-coated flexible polyester sheets have a hard, abrasion resistant coating and solvent resistant substrate for rapid, clean separations....
Supplier: VWR Collection
Description: These TLC glass plates are suitable for easy and quick sample applications.
Sale
Supplier: VWR Collection
Description: These TLC aluminium plates are suitable for easy and quick sample applications. They are flexible and can be cut to match different plate size require...
Supplier: VWR Collection
Description: These HPTLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.
Supplier: VWR Collection
Description: These HPTLC glass plates use an optimised particle size which results in better separation efficiency.
Supplier: VWR Collection
Description: These TLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.
Supplier: Avantor
Description: Thin-layer chromatography plates (TLC), J.T.Baker®, Coating material: Silica gel
Supplier: VWR Collection
Description: Modified silica layers are, depending on the modification, suitable for polar to unpolar eluents or eluent mixtures.
Supplier: Avantor
Description: Ready-to-use in thin layer chromatography applications, Baker-flex® precoated flexible plastic sheet have a hard, abrasion-resistant coating and solve...
Catalog Number: (552-0217)
Supplier: MACHEREY-NAGEL
Description: Glass plates with mixed layers for TLC.
UOM: 1 * 25 items
Supplier: MACHEREY-NAGEL
Description: Thin-layer chromatography plates (TLC), POLYGRAM®, POLYGRAM® TLC polyester sheets Alox N/UV254, Coating material: Aluminium oxide (Alox), Backing material: Polyester, Fluorescent indicator: F254, Plate size: 40×80 mm
Supplier: MACHEREY-NAGEL
Description: Unmodified HPTLC silica layers available as glass plates with or without fluorescent indicator. The narrow fractionation of the silica particles featu...
Supplier: MACHEREY-NAGEL
Description: Amino-modified HPTLC silica layers are available as glass plates or ALUGRAM® aluminium sheets. They are ideal for the separation of vitamins, sugars, ...
Supplier: MACHEREY-NAGEL
Description: Unmodified standard silica layers for TLC.
Supplier: MACHEREY-NAGEL
Description: These TLC plates and sheets are prepared by hydrolysis of high purity cellulose with HCl. They are used for carboxylic acids, lower alcohols, urea and...
Supplier: MACHEREY-NAGEL
Description: Unmodified HPTLC silica layers for HPTLC.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom