You Searched For: Precolumn Filters and Frits
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: These cap style frits used to preserve the packaging material within the column itself.
Supplier: Avantor
Description: These precolumn filters with PEEK body and stainless steel frit provide protection for analytical and microbore columns. They connect directly to the ...
Supplier: Avantor
Description: Frit made of 316 grade stainless steel.
Supplier: Avantor
Description: Placed immediately before the column, the precolumn filter traps particles introduced with the sample. For samples, which may irreversibly absorb onto...
Catalog Number: (HICHHI-140)
Supplier: Avantor
Description: Avantor® Hichrom, Preparative frit assembly, Natural, Porosity: 2 µm
UOM: 1 * 10 items
Supplier: Avantor
Description: Avantor® ACE® UHPLC pre-column filters made from stainless steel/PEEK provide protection for all UHPLC columns. They offer ultra-low dispersion for ma...
Supplier: Avantor
Description: Avantor® Hichrom, Frits, Black, Porosity: 0,5 µm, Frit material: Stainless steel
Catalog Number: (7121-05)
Supplier: Avantor
Description: J.T.Baker® BAKERBOND spe™, Frits for 3 ml polypropylene columns
UOM: 1 * 50 items
Supplier: Avantor
Description: J.T.Baker® BAKERBOND®, PTFE frits for 3 ml glass columns
Catalog Number: (HI-300)
Supplier: Avantor
Description: The frit removal tool is ideal for the removal of frits used in HPLC/LC columns.
UOM: 1 * 1 items
Catalog Number: (HICHHI-1140)
Supplier: Avantor
Description: Avantor® Hichrom, Frit, Porosity: 2 µm, Frit material: PEEK
UOM: 1 * 1 items
Supplier: Bohlender
Description: These frits are made from PTFE.
Supplier: Bohlender
Description: These PTFE gas distributors and frits are suitable for steady sparkling of the flowing gas.
Supplier: MACHEREY-NAGEL
Description: CHROMABOND®, CHROMABOND® Flash DL 120, PE filter elements, 36,0 mm ID
Catalog Number: (MICS49650-005)
Supplier: MICROSOLV TECH CORP
Description: The flter protects columns from particulates and extends the column life-time.
UOM: 1 * 10 items
Supplier: Restek
Description: These 6 mm frits are manufactured from fine, chromatographic grade 316 stainless steel and different sizes are available to fit most column and pore s...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom