Ampoules, pre-scored, KIMBLE®
Supplier: DWK Life Sciences
KIML12010L-1EA 0 EUR
KIML12010L-1 KIML12040G-2 KIML12010L-2 KIML12010L-5
Ampoules, pre-scored, KIMBLE®
Ampoules
These ampoules made from borosilicate glass, Type I are hermetically sealed and provide maximum solution stability.
  • Stems are pull sealed
  • Excellent dimensional stability for high speed filling
  • Amber glass is for light sensitive compounds
The pre-scored constricted portion of the stem permits safe and easy opening while eliminating the need for filing.