You Searched For: Fraction Collectors
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GILSON LCMS
Description: Ideal solutions for small to large capacity collection. Gilson offers advanced solutions for fraction collection with a complete line of stand‑alone a...
Supplier: COLLECTION PREFIX HPLC
Description: Foxy R1 and Foxy R2 fraction collectors are ideal for a lot of applications including HPLC, preparative HPLC, protein and peptide purification. The Fo...
Catalog Number: (29-0230-51)
Supplier: Cytiva
Description: Frac30 is a round fraction collector designed for use with the ÄKTA start purification system. The Frac30 is controlled via the ÄKTA start touch scree...
UOM: 1 * 1 items
Catalog Number: (905-0125)
Supplier: LABOMATIC INSTRUMENTS
Description: The Vario 4000 fraction collector is an open, height-adjustable system without limits on the number of fractions, and is compatible with a wide range ...
UOM: 1 * 1 items
Catalog Number: (HARVCMA8002770)
Supplier: HARVARD APPARATUS LTD
Description: The CMA 470 refrigerated fraction collector is specifically designed to collect microliter volume fractions typical of microdialysates. It is a stand ...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom