Precast horizontal gel kits, SERVALYT™ PRECOTES™ PGM Kit
: Serva
For PGM analysis by isoelectric focusing.
SERVALYT™ PRECOTES™ Hp Kit contains:
5× SERVALYT™ PRECOTES™ Hp gels, 245×125 mm or 125×125 mm, 300 µm
Anode and cathode buffer solutions
Applicator strips and electrode wicks
- SeqPk
- SERVALYT™ PRECOTES™ PGM kit, 245×125 mm
Specifications
Environmentally Preferable - SERVALYT™ PRECOTES™ PGM kit, 125×125 mm