Reusable earplugs, E-A-R™ Ultrafit™ 14
Supplier: 3M
E-A-R™ UltraFit™
MMMAUF01015EA 0 EUR
MMMAUF01015
Reusable earplugs, E-A-R™ Ultrafit™ 14
Earplugs Reusable Earplugs
These pre-moulded earplugs made from soft TPE use unique filter technology to take the edge off low levels of harmful noise whilst allowing normal conversations or listening to broadcast music.
  • Moisture resistant and compatible with other PPE
  • No need for any roll down
  • Triple-flanged tip, pistol grip stem and angled shape for easy insertion
  • Washable and reusable
Recommended for noise levels up to 85 dB(A).
SNR: 14 dB
Certifikace: EN 352-2
Upozornení: * One size fits all.