Brightfield cell counters, Cellometer®
Supplier: Nexcelom Bioscience
Cellometer®
NEXCCMAUTOT4PLUSEA 5000 EUR
NEXCCMAUTOT4PLUS NEXCCMAT4-SK150
Brightfield cell counters, Cellometer®
Cell Counters and Flow Cytometers Automated Cell Counters
These automated cell counters are designed for counting small cells.
  • Highly reproducible results can be saved and exported in Excel
  • Improved data quality, versatility and flexibility
  • Templates for hundreds of cell types and easy definition of types of cells
  • Brightfield technique, powerful optics, pointer and analysis software
Applications usually include insect, algae, yeast, cancer, blood cells and spores.
Delivery information: Supplied with a box of counting chambers.