Slovenia
Order Entry
Anti-HA tag Rabbit Polyclonal Antibody (DyLight® 650)
  COBSD10-1830
Anti-HA tag Rabbit Polyclonal Antibody (DyLight® 650)
  COBSD10-1830
 
629,00 €
   
  1 * 100 µG

 

  • Antibody type:
    Tag
  • Antigen name:
    HA tag
  • Antigen symbol:
    HA
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: