Anti-SRI Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS5363-01BEA 633 EUR
USBIS5363-01B
Anti-SRI Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: SRI (sorcin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR