Anti-SRI Mouse Monoclonal Antibody [clone: 11C791]

Supplier: United States Biological
USBIS5363-01EA 522 EUR
USBIS5363-01
Anti-SRI Mouse Monoclonal Antibody [clone: 11C791]
Antibodies
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: SRI (sorcin)
Clonality: Monoclonal
Clone: 11C791
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR