Slovenia
Order Entry
Anti-ACAD10 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-ACAD10 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB125378-100
  • BIRBORB125378-200
  • Antigen symbol:
  • ACAD10
  • ACAD10
  • Antigen name:
  • acyl-CoA dehydrogenase family, member 10
  • acyl-CoA dehydrogenase family, member 10
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • ImmunoChemistry:
  • Yes
  • Yes
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • BIRBORB125378-100
    ORB125378-100
    100 µg
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG - 0,00 €
    0,00 €
    1 * 100 µG
       0,00 €

     
     
    Specifications
    Antigen symbol
    ACAD10
    Antigen name
    acyl-CoA dehydrogenase family, member 10
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    ImmunoChemistry
    Yes
    Purification
    purified by affinity chromatography
    Concentration
    1 mg/ml
    Storage buffer
    PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB125378-200
    ORB125378-200
    200 µg
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG - 0,00 €
    0,00 €
    1 * 200 µG
       0,00 €

     
     
    Specifications
    Antigen symbol
    ACAD10
    Antigen name
    acyl-CoA dehydrogenase family, member 10
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    ImmunoChemistry
    Yes
    Purification
    purified by affinity chromatography
    Concentration
    1 mg/ml
    Storage buffer
    PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
    Storage temperature
    Cool
    Shipping temperature
    Ice