Nitrile gloves, Microflex® 93-853
: Ansell
- Catalog No:
- 112-5159
- 112-4278
- 112-4275
- 112-4279
- 112-4276
- 112-5160
- 112-4277
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- SeqGlove sizeColourLengthThicknessPackedPk
- 112-427593-853/5.5-6015,5 - 6Blue295 mm0,14 mm50/box501 * 50 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+ - 112-427693-853/6.5-7026,5 - 7Blue295 mm0,14 mm50/box501 * 50 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+ - 112-427793-853/7.5-8037,5 - 8Blue295 mm0,14 mm50/box501 * 50 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+ - 112-427893-853/8.5-9048,5 - 9Blue295 mm0,14 mm50/box501 * 50 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+ - 112-427993-853/9.5-10059,5 - 10Blue295 mm0,14 mm50/box501 * 50 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+ - 112-515993-853/10.5-110610,5 - 11Blue295 mm0,14 mm50/box501 * 50 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+ - 112-516093-853/11.5-120711,5 - 12Blue295 mm0,14 mm40/box401 * 40 items25,90 €
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingertipsCuff styleBeadedEN normEN 455, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUSAnti-static+