You Searched For: ID Card Holders
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (PRTWID10YER)
Supplier: Portwest
Description: In Case of Emergency (ICE) stickers for accident victims gives on-site instant access to victims potential lifesaving information and enables emergenc...
UOM: 1 * 1 items
Catalog Number: (KWIN100550940)
Supplier: FRISTADS KANSAS GROUP
Description: ID-card holder, plastic, attached with clip or snap hook, pack of 10, Colour: Black
UOM: 1 * 10 items
Catalog Number: (DURA832701)
Supplier: DURABLE
Description: ID card holder with clasp type metal carabiner and 800 mm cable.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 3 of 3
no targeter for Bottom