Peqlab PerfectBlue Maxi S Plus, Gel system
Supplier: Avantor
Peqlab PerfectBlue Maxi S Plus
700-0538EA 991 EUR
700-0538
Peqlab PerfectBlue Maxi S Plus, Gel system
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The smallest of the Maxi series, the Maxi S Plus is suitable for high quality sample separation of up to 160 samples per run. With this flexible system, the gel size can be split into two, three or four sections by using different comb positions, plus the gel size can be reduced using the wall comb or the casting dam system. The system is supplied with three combs but others are available - either standard combs, or microtitre combs which are ideal for use with multichannel pipettes. Use of the system end gates ensures that gel pouring is leak-free and easy. The levelling table keeps the resulting gel flat, maximising the quality of the run.
*Based on an agarose gel of approximately 5 mm thickness
Informace pro objednávání: Supplied with three combs (1,5 mm thickness, 10, 17, 34 teeth), lid, gel tray, three point levelling platform, tank, cables and end gates.