These data loggers are designed to measure wide temperature ranges (down to −200 °C) in laboratories or during transit.
- Recording interval
- Start delay, time zone, stop key enable
- Alarm set points: Ambient and external probe individually
- Cumulative alarm duration from user-selected minimum/maximum temperature values
- Reusable/re-programmable for monitoring cryo material in transit as well as reagents, clinical samples, frozen samples, pharmaceuticals and food items
External stainless steel probe (L×Ø: 110×3,5 mm) with PTFE cable
* Ambient
** Probe
Certifikace: Certification compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Starter kit includes Traceable® Logger-Trac™ LN2 logger, data capture cable, MaxiThermal software package, battery and Traceable® certificate of calibration.