Order Entry
Amino acid F for Rapid liquid silica method
 :  Hach
undefined
Amino acid F for Rapid liquid silica method
 :  Hach
  • Environmentally Preferable:
  •  
    Seq
    Pk
  • Amino acid F reagent powder, for rapid liquid silica, 55 g
    Amino acid F reagent powder, for rapid liquid silica, 55 g
    HACH2651155
    2651155
    01
    55 g
    1 * 55 g
    1 * 55 g - 52,00 €
    52,00 €
    1 * 55 g
       52,00 €
  • Amino acid F dilution solvent, for rapid liquid silica, 475 ml
    Amino acid F dilution solvent, for rapid liquid silica, 475 ml
    HACH2353011
    2353011
    02
    475 mL
    1 * 475 mL
    1 * 475 mL - 40,10 €
    40,10 €
    1 * 475 mL
       40,10 €