Electronic flow meter
Supplier: PerkinElmer
PerkinElmer®
PERCN9307029EA 1960 EUR
PERCN9307029
Electronic flow meter
Flow Meters
This portable flow meter allows for real-time flow measurements.
  • Quick split ratio mode calculation
  • Ratios are displayed in real time
  • LCD alphanumeric display
Operating temperature from 0 to 45 °C.
Certifikace: Traceable to the National Institute of Standards and Technology (NIST).
Upozornení: It is not compatible with corrosive or flammable gases.