PCR storage racks
211-0445EA 0 EUR
VWRI211-0445
PCR storage racks
PP.
  • These racks will hold tubes, strips, or plates
  • Separate snap on lid
  • Autoclavable and freezable
The 96-well PCR storage rack is perfect for storing samples before cycling or working with completed procedures.
Informace pro objednávání: Available in an assortment pack of five colours: red, orange, yellow, green and blue.