Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (1)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "scrub-brushes"
https://si.vwr.com/stibo/smallweb/std.lang.all/96/44/18709644.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
Supplier:  PFENNIG REINIGUNGSTECHNIK
Description:   Joint scrubber/brush for gaps in hygienically sensitive areas.
Catalog Number: (PPSA3590500)
Supplier:  PFENNIG REINIGUNGSTECHNIK
Description:   Hand brush with long handle for hygienically sensitive areas.
UOM:  null
Catalog Number: (MICRBRWH-30)
Supplier:  MICRONOVA
Description:   Brush, For: Valve, Valve brush, autoclavable, Material: Nylon bristle, Head Ø: 102 mm, Total length: 762 mm, Handle: Plastic handle
UOM:  1 * 1 items
Catalog Number: (VILE145873)
Supplier:  FHP VILEDA SCS
Description:   Brush in strong density polypropylene plastic stock incl. pin hole and high quality polyester PBT-filaments
UOM:  1 * 1 items
Catalog Number: (KINGVI00392)
Supplier:  KING DISPOSABLES
Description:   These brushes include plastic frame with white and blue PBT bristles.
UOM:  1 * 10 items
Supplier:  GORDON BRUSH MFG CO
Description:   This traditional round brush is made of Pure Red Sable hair, which is softer than camel hair, pony hair and bristle. Ideal for applying oils, acrylics...
Catalog Number: (NEOL2-3275)
Supplier:  NEOLAB
Description:   Brush, Scrubbing brush, without handle, Total length: 220 mm
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7  of 7