Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (60)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "labware-cleaning-brushes"
https://si-bot.vwr.com/stibo/smallweb/std.lang.all/53/23/18945323.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"38"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (149-0134)
Supplier:  VWR Collection
Description:   Brushes with white nylon bristles and wooden handle. Ideal for intensive, extra-thorough cleaning of glass beakers and a wide range of other wide-nec...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   Brushes with white or black nylon, with wire handle. These cleaning brushes are specially designed for cleaning gas washing bottles, cylinders, and la...
Supplier:  VWR Collection
Description:   Brushes with white nylon or natural bristles and wire handle. Ideal for cleaning reagent flasks, burettes, pipettes, tubes and a wide variety of other...
Supplier:  VWR Collection
Description:   Brushes with white nylon bristles with wire handle. Ideal for cleaning volumetric pipettes, milk pipettes, and Mohr pipettes.
Supplier:  VWR Collection
Description:   Brushes with white, natural bristles and flexible plastic handle. Ideal for cleaning Erlenmeyer flasks, round and flat bottom flasks, Kjeldahl flasks,...
Catalog Number: (149-0138)
Supplier:  VWR Collection
Description:   Natural black bristles with wire handle
UOM:  1 * 5 items
Supplier:  VWR Collection
Description:   Labware cleaning brush, For: Tubes, Material: White nylon bristles, Head Ø: 13 mm, Trimming length: 78 mm, Total length: 200 mm, Handle: Wire handle
Supplier:  VWR Collection
Description:   Brushes with white nylon bristles and wire handle. These brushes are specially designed for cleaning small tubes, pipes, and a wide range of small con...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 38
  1  2  3  4  5  Next