You Searched For: Wafer Carriers
26  results were found
SearchResultCount:"26"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: ENTEGRIS INC
Description: Holds one mask or reticle.
Supplier: ENTEGRIS INC
Description: 182 Series
The industry standard 150 mm process wafer carrier. Four-point contact on the 'H' bar for improved wafer positioning. Open sidewall d...
Supplier: ENTEGRIS INC
Description: These wafer baskets made of PFA are designed for processing and handling substrates. They are compatible with tanks and handles.
Supplier: ENTEGRIS INC
Description: These carriers are designed to hold masks or reticles during transport. They can be used within diverse industries, such as semi-conductor data storag...
Supplier: ENTEGRIS INC
Description: Squeeze style.
Supplier: ENTEGRIS INC
Description: These standard 150 mm wafer carriers feature a high rigidity and dimensional stability.
Supplier: ENTEGRIS INC
Description: These 100 mm wafer carriers are designed for optimum performance in automated operations or for processing LCDs and other small devices.
Sale
Supplier: ENTEGRIS INC
Description: Provide excellent security for masks and reticles with pellicles.
Sale
Supplier: ENTEGRIS INC
Description: Squeeze style
Attaches to the notches on both end walls of carrier
Overall size: 163×140×183 mm
Compatible with all 'M' style carriers
Supplier: ENTEGRIS INC
Description: These carriers are designed to hold masks or reticles during transport. Carriers and boxes purchased as an assembly.
Supplier: ENTEGRIS INC
Description: 2,5" wafer carriers, Pocket spacing: 4,76 mm, pocket flat: 1,27 mm, 25 wafers, PFA, Natural
Supplier: ENTEGRIS INC
Description: These wafer warrier handles are designed for extended lead times apply.
Sale
Supplier: ENTEGRIS INC
Description: These standard 125 mm wafer carriers are designed for optimum performance in automated equipment.
Supplier: ENTEGRIS INC
Description: Provides excellent security for masks or reticles with pellicles.
Supplier: ENTEGRIS INC
Description: Squeeze style handles.
Supplier: ENTEGRIS INC
Description: PFA, natural.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 26
no targeter for Bottom