You Searched For: Wafer Storage Boxes
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: ENTEGRIS INC
Description: E 99 Series
Sale
Supplier: ENTEGRIS INC
Description: Wafer handling, Wafer storage boxes for 76,2 mm (3") wafer handling, E 52-101 series, Storage box, PP, Blue
Supplier: ENTEGRIS INC
Description: With pin and socket hinges. Single snap latch.
Sale
Supplier: ENTEGRIS INC
Description: Spring boxes, Holds five trays and one cover or six trays, STAT-PRO® 125
Supplier: ENTEGRIS INC
Description: Two snap latches, one swing pin latch or three swing pin latches. Pin and socket hinges.
Sale
Supplier: ENTEGRIS INC
Description: E 90 series
Catalog Number: (ENTEE04-1202)
Supplier: ENTEGRIS INC
Description: Wafer handling, Wafer storage boxes for 2,5" and smaller wafer handling, E 04 series, Polystyrene, Black
UOM: 1 * 4 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom