Order Entry
General purpose gloves, AlphaTec® 87-195
General purpose gloves, AlphaTec® 87-195
Supplier:  Ansell
Certificates
undefined
General purpose gloves, AlphaTec® 87-195
Supplier:  Ansell
Specifications
  • Catalog No:
  • 112-6437
  • 112-6438
  • 112-6439
  • 112-6440
  • Model:
  • 87-195
  • 87-195
  • 87-195
  • 87-195
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Cotton
  • Finish:
  • Fishscale
  • Fishscale
  • Fishscale
  • Fishscale
  • Cuff style:
  • Straight
  • Straight
  • Straight
  • Straight
  • EN norm:
  • EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
  • EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
  • EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
  • EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
  • Food contact approved:
  • +
  • +
  • +
  • +
Specifications
Product Information
  • Catalog #
    Supplier #
    Seq
    Glove size
    Colour
    Length
    Thickness
    Pk
    Availability
    Price
  • 112-6437
    87-195/6.5-7
    01
    6,5 - 7
    Blue
    305 mm
    0,35 mm
    12 PAIR
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 21,20 €
    21,20 €
    New Customer Promotional Price
    1 * 12 PAIR
       21,20 €
    Specifications:
    More
    Specifications
    Model
    87-195
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Finish
    Fishscale
    Cuff style
    Straight
    EN norm
    EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
    Food contact approved
    +
  • 112-6438
    87-195/7.5-8
    02
    7,5 - 8
    Blue
    305 mm
    0,35 mm
    12 PAIR
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 21,20 €
    21,20 €
    New Customer Promotional Price
    1 * 12 PAIR
       21,20 €
    Specifications:
    More
    Specifications
    Model
    87-195
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Finish
    Fishscale
    Cuff style
    Straight
    EN norm
    EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
    Food contact approved
    +
  • 112-6439
    87-195/8.5-9
    03
    8,5 - 9
    Blue
    305 mm
    0,35 mm
    12 PAIR
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 21,20 €
    21,20 €
    New Customer Promotional Price
    1 * 12 PAIR
       21,20 €
    Specifications:
    More
    Specifications
    Model
    87-195
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Finish
    Fishscale
    Cuff style
    Straight
    EN norm
    EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
    Food contact approved
    +
  • 112-6440
    87-195/9.5-10
    04
    9,5 - 10
    Blue
    305 mm
    0,35 mm
    12 PAIR
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 21,20 €
    21,20 €
    New Customer Promotional Price
    1 * 12 PAIR
       21,20 €
    Specifications:
    More
    Specifications
    Model
    87-195
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Finish
    Fishscale
    Cuff style
    Straight
    EN norm
    EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
    Food contact approved
    +