VWR® B'mat Decontamination Flooring
Supplier: Avantor
VWR® B´Mat VWR® B'mat
115-1846EA 0 EUR
115-1846
VWR® B'mat Decontamination Flooring
Floor Mats Permanent Floor Mats
The decontamination flooring is made from proprietary polymer with a fabric backing. Particles are naturally attracted by the flooring surface, and are released only by a wet cleaning. To restore its full efficacy, the flooring only has to be mopped, rinsed and dryed. The core properties of the polymer (bacteriostatic, ESD, non outgassing) make of the b'mat flooring the ideal solution for contamination control in gowning areas, in material entry zones, and in cleanrooms.
  • Attracts and retains over 98% of foot-borne and wheel-borne particles
  • With a low profile (only 2,5 mm thick), and thanks to beveled PVC edges welded on all sides, operators can walk without tripping and carts can be pushed without effort
  • Flooring requires to be glued to the floor, which ensures a perfect hold and a maximum lifetime
  • Due to its exceptional weight resistance, the b'mat flooring is compatible with foot traffic, but also with heavy lift- or hand-trucks
  • Good chemical resistance and heat resistance between 0 and 50 °C
Colour: Grey