You Searched For: Gel Skin Disinfectants
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SCHUELKE & MAYR GMBH
Description: Alcohol-based hand rub for hygienic and surgical hand disinfection with panthenol and vitamin E, without colour and perfume.
Catalog Number: (BODL9801440)
Supplier: Hartmann Bode
Description: Baktolan® Balm is a water-in-oil balm with high quality nourishing components that were particularly adapted to the skin’s pH value (5,5). This stabil...
UOM: 1 * 350 mL
Supplier: PHAGOGENE
Description: Hydroalcoholic gels and solutions for the disinfection of the hands. Special formulation enriched with moisturisers and emollients, ideal for sensitiv...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 3 of 3
no targeter for Bottom