You Searched For: Hand and Body Lotions
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Deb-STOKO
Description: Soft cream (O/W emulsion) with moisturising effect for the effective care of normal skin. Suitable for all over body use. Suitable for people prone to...
Supplier: Deb-STOKO
Description: Deb stoko sanitise gel dispenser can be used in office and facility, industrial, food, hospitality and education sectors.
Catalog Number: (115-2354)
Supplier: Deb-STOKO
Description: This foot spray helps to protect feet by preventing excessive perspiration, for instance when wearing closed in footwear, such as safety shoes, rubber...
UOM: 1 * 25 items
Supplier: Deb-STOKO
Description: Softcream (O/W emulsion) with moisturising effect for the effective care of normal skin. Suitable for all over body use and for use under latex and ni...
Supplier: Deb-STOKO
Description: Perfume-free, water resistant 5-star rated, factor 50 sunscreen prolongs the skin's natural protection time against UVA, UVB and UVC exposure. For use...
Supplier: Deb-STOKO
Description: Stokolan® classic is a concentrated moisturising and conditioning cream that helps to restore moisture in normal and dry skin. Ideal for use in indust...
Supplier: Deb-STOKO
Description: Stokolan® Intense is a nurturing, regenerative and barrier active creamy lotion containing grapeseed oil, ceramid complex, allantoin, ginkgo biloba, u...
Supplier: Diversey
Description: Cleans and disinfects hands in one easy step. Based on lactic acid, an organic based ingredient from Natural sources that inhibits the growth of micro...
Safer Human and Environmental Health
Sustainable Materials
Supplier: Diversey
Description: Gentle and effective hand wash with a 100% natural perfume. Cleans hands leaving them hygienically fresh.
Catalog Number: (FAWE13042001)
Supplier: FAWECO HAUTSCHUTZ
Description: Ligana® Multi-tec is a skin protection lotion with dual effect, which is used when handling varying workplace substances. It does not leave any annoyi...
UOM: 1 * 10 Bottle
Catalog Number: (115-1364)
Supplier: FAWECO HAUTSCHUTZ
Description: Barrier cream against natural and artificial UV rays, e.g. for working outdoors or in electric welding. Particularly suitable for highly sensitive ski...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom