Order Entry
Sample dipper with v bottom
Sample dipper with v bottom
 :  ENTEGRIS
Sample dipper with v bottom
 :  ENTEGRIS
  •  
    Pk
  • Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
    Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
    ENTED11-0215
    D11-0215
    4
    1 * 4 items
    1 * 4 items
    Specifications
    Environmentally Preferable
  • Dipper, wafer size 57 - 125 mm, square plate size 50 - 153 mm, max. thickness 3,0 mm
    Dipper, wafer size 57 - 125 mm, square plate size 50 - 153 mm, max. thickness 3,0 mm
    ENTED14-0215
    D14-0215
    4
    1 * 4 items
    1 * 4 items
    Specifications
    Environmentally Preferable
  • Dipper, wafer size 125 - 200 mm, square plate size 114 - 203 mm, max. thickness 6,4 mm
    Dipper, wafer size 125 - 200 mm, square plate size 114 - 203 mm, max. thickness 6,4 mm
    ENTED15-0215
    D15-0215
    4
    1 * 4 items
    1 * 4 items