You Searched For: Drying Tubes
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors with 6 mm external diameter.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Clear DURAN® borosilicate 3.3 glass. Angled at 75° with ground joint cone.
Catalog Number: (201-9110)
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear. One bulb and glass tubing connection.
UOM: 1 * 1 items
Supplier: KARTELL
Description: PE.
Supplier: DWK Life Sciences
Description: DURAN® borosilicate glass 3.3. clear. Angled at 75° with ground joint cone.
Supplier: Bel-Art Products, a Part of SP
Description: Unbreakable polyethylene straight tubes.
Supplier: WITEG LABORTECHNIK GMBH
Description: Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.
Supplier: WITEG LABORTECHNIK GMBH
Description: Borosilicate glass 3.3. Angled at 75° with ground joint cone.
Supplier: SciLabware
Description: PYREX®, borosilicate glass with ground glass cone.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom