Single channel micro pipettes, electronic, variable / fixed volume, Accumax VA-FA
Supplier: Accumax
PFACVA-100EA 0 EUR
FINEFA-0020 PFACVA-100 FINEFA-0050 PFACVA-300 FINEFA-0500 PFACVA-200 FINEFA-0005 PFACVA-500 PFACVA-400 PFACVA-700 FINEFA-0100 PFACVA-600 PFACVA-900 FINEFA-0025 PFACVA-800 PFACFA-2000 PFACFA-1000 PFACFA-5000
Single channel micro pipettes, electronic, variable / fixed volume, Accumax VA-FA
Pipettes Micropipettes
These pipettors feature a click-stop digital system for volume setting which ensures accurate dispensing of the liquid. The two-step plunger operation allows a reverse pipetting technique.
  • Smooth plunger mechanism for ease of operation
  • Tip ejector for smooth ejection of the tip preventing any contamination
  • Ergonomic, lightweight design with comfortable finger rest
  • Durable tip cone offers high resistance to shocks and chemical corrosion
  • Colour-coded for easy identification
  • Autoclavable (lower part)
Informace pro objednávání: Supplied with calibration report.