Commercial fridge/freezer, GCV4060
Supplier: Liebherr
471-1113EA 2259 EUR
471-1113
Commercial fridge/freezer, GCV4060
Refrigerators
This cabinet is ideal for use in a restricted space.
  • Moulded polystyrol interior (for ease of cleaning and improved hygiene)
  • Tropicalised to perform in ambient temperatures of up to 43 °C
  • Dual controllers and dual compressors for optimum performance
  • Electronic controller
  • Static cooling system
Temperature range: +1 to 15 °C for the fridge and −14 to −28 °C for the freezer