Surface sanitiser wipes, IW50
Supplier: Portwest
PRTWIW50BLUEA 10 EUR
PRTWIW50BLU
Surface sanitiser wipes, IW50
Wipes Disposable Wipes and Towels Disinfectant Wipes
Anti-bacterial polyviscose wipes designed to protect surfaces and prevent contamination
  • Practical dispenser box for single sheet dispensing
  • Ideal for sanitising surfaces prior to food preparation
Certifikace: EN 1276, EN 14476
Upozornení: Use biocides safely. Always read the label and product information before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904