Water treatment systems
Supplier: smeg
171-0851EA 3160 EUR
171-0851 171-0849 171-0850
Water treatment systems
Water Purification Systems Deionisers
The WSE series of appliances are technological water softeners for GW3060, GW4090 and GW6090 capable of completely eliminating lime from water. The resins are regenerated using normal sodium chloride. Suitable for softening WD6090 glassware washer supply water and for centralised treatment systems. Available in versions with flow rates of up to 2700 l/H. They provide backflow regeneration based on the volume of water (bearing in mind the resin saturation rate) and proportional brine in order to optimise water and salt consumption.
  • Fitted with an electronically controlled head which can be programmed depending on the hardness of the water
  • Integrated salt tank