Order Entry
Standard duty piston pumps, WOB-L
Standard duty piston pumps, WOB-L
undefined
Standard duty piston pumps, WOB-L
  • Catalog No:
  • ILMV2561C-50
  • ILMV2546C-02
  • ILMV2534C-02
  • Model:
  • WOB-L 2561
  • WOB-L 2546
  • WOB-L 2534
  • Electrical supply:
  • 250 W
  • 190 W
  • 93 W
  • Weight:
  • 7,5 kg
  • 5,3 kg
  • 5,3 kg
  •  
    Seq
    Flow rate
    Pk
  • Standard duty piston pump, dry, WOB-L 2534, 230 V
    Standard duty piston pump, dry, WOB-L 2534, 230 V
    ILMV2534C-02
    2534C-02
    02
    28 l/min at 50 Hz
    34 l/min at 60 Hz
    1
    1 * 1 items
    1 * 1 items - 675,00 €
    675,00 €
    1 * 1 items
       675,00 €
    Accessories
    Specifications
    Model
    WOB-L 2534
    Environmentally Preferable
    Electrical supply
    93 W
    Weight
    5,3 kg
  • Standard duty piston pump, dry, WOB-L 2546, 230 V
    Standard duty piston pump, dry, WOB-L 2546, 230 V
    ILMV2546C-02
    2546C-02
    03
    38 l/min at 50 Hz
    45 l/min at 60 Hz
    1
    1 * 1 items
    1 * 1 items - 825,00 €
    825,00 €
    1 * 1 items
       825,00 €
    Accessories
    Specifications
    Model
    WOB-L 2546
    Environmentally Preferable
    Electrical supply
    190 W
    Weight
    5,3 kg
  • Standard duty piston pump, dry, WOB-L 2561, 230 V
    Standard duty piston pump, dry, WOB-L 2561, 230 V
    ILMV2561C-50
    2561C-50
    04
    50 l/min at 50 Hz
    60 l/min at 60 Hz
    1
    1 * 1 items
    1 * 1 items - 1 595,00 €
    1 595,00 €
    1 * 1 items
       1 595,00 €
    Accessories
    Specifications
    Model
    WOB-L 2561
    Environmentally Preferable
    Electrical supply
    250 W
    Weight
    7,5 kg