You Searched For: Flow Cells
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (662-0457)
Supplier: VWR Collection
Description: VWR® low flow cells are designed for use in a variety of instrumental techniques such as FIA, HPLC and IEF. They are also used to enable a continuous ...
UOM: 1 * 1 items
Supplier: Oxford Nanopore Technologies
Description: Nanopore sequencing flow cells for use with MinION or MinION Mk1C devices.
Supplier: Oxford Nanopore Technologies
Description: Flongle is an adapter for MinION devices that enables direct, real-time DNA sequencing on smaller, single-use Flongle Flow Cells.
Catalog Number: (METT51302257)
Supplier: Mettler - Toledo
Description: Flow through cell, glass, for electrodes with 12 mm diameter
UOM: 1 * 1 items
Catalog Number: (METT30014098)
Supplier: Mettler - Toledo
Description: Flow cell, InLab® Trace
UOM: 1 * 1 items
Supplier: Agilent
Description: Quartz body, 0,3 mm
Catalog Number: (HEWLG4301-60100)
Supplier: Agilent
Description: Flow cell, high pressure, flow path 10 mm, for supercritical fluid chromatography
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom