Flow cups with fixed nozzle
- SeqRangeCup orificePk
- Viscosity flow cup (max.W×H = 92×84 mm)Viscosity flow cup (max.W×H = 92×84 mm)769-1152VF20480017 - 42 cSt3 mm11 * 1 items191,29 €
Accessories
Specifications
Environmentally Preferable - Viscosity flow cup (max.W×H = 92×84 mm)Viscosity flow cup (max.W×H = 92×84 mm)SIMEVF2049VF20490234 - 135 cSt4 mm11 * 1 items191,29 €
Accessories
Specifications
Environmentally Preferable - Viscosity flow cup (max.W×H = 92×84 mm)Viscosity flow cup (max.W×H = 92×84 mm)SIMEVF2183VF21830391 - 326 cSt5 mm11 * 1 items212,28 €
Accessories
Specifications
Environmentally Preferable - Viscosity flow cup (max.W×H = 92×84 mm)Viscosity flow cup (max.W×H = 92×84 mm)SIMEVF2050VF205004188 - 684 cSt6 mm11 * 1 items191,29 €
Accessories
Specifications
Environmentally Preferable - Viscosity flow cup (max.W×H = 92×84 mm)Viscosity flow cup (max.W×H = 92×84 mm)SIMEVF2051VF205105600 - 2000 cSt8 mm11 * 1 items212,28 €