Order Entry
Moisture traps, MT series
Moisture traps, MT series
Supplier:  Agilent
Certificates
Moisture traps, MT series
Supplier:  Agilent
Specifications
  • Catalog No:
  • HEWLMT120-2
  • HEWLMT200-2
  • HEWLMT400-2
  • HEWLMT120-4
  • HEWLMT200-4
  • Fitting size:
  • 1/8"
  • 1/8"
  • 1/4"
  • 1/4"
  • 1/4"
Specifications
Product Information
  • Catalog #
    Supplier #
    Seq
    Pk
    Availability
    Price
  • Refillable moisture trap, 120 ml, removal capacity 21,6 g, max. effluent H₂O concentration 20 ppb
    Refillable moisture trap, 120 ml, removal capacity 21,6 g, max. effluent H₂O concentration 20 ppb
    HEWLMT120-2
    MT120-2
    01
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 240,00 €
    240,00 €
    New Customer Promotional Price
    1 * 1 items
       240,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Fitting size
    1/8"
  • Refillable moisture trap, 120 ml, removal capacity 21,6 g, max. effluent H₂O concentration 20 ppb
    Refillable moisture trap, 120 ml, removal capacity 21,6 g, max. effluent H₂O concentration 20 ppb
    HEWLMT120-4
    MT120-4
    02
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 236,00 €
    236,00 €
    New Customer Promotional Price
    1 * 1 items
       236,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Fitting size
    1/4"
  • Refillable moisture trap, 200 ml, removal capacity 36 g, max. effluent H₂O concentration 18 ppb
    Refillable moisture trap, 200 ml, removal capacity 36 g, max. effluent H₂O concentration 18 ppb
    HEWLMT200-2
    MT200-2
    03
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 250,00 €
    250,00 €
    New Customer Promotional Price
    1 * 1 items
       250,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Fitting size
    1/8"
  • Refillable moisture trap, 200 ml, removal capacity 36 g, max. effluent H₂O concentration 18 ppb
    Refillable moisture trap, 200 ml, removal capacity 36 g, max. effluent H₂O concentration 18 ppb
    HEWLMT200-4
    MT200-4
    04
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 250,00 €
    250,00 €
    New Customer Promotional Price
    1 * 1 items
       250,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Fitting size
    1/4"
  • Refillable moisture trap, 400 ml, removal capacity 72 g, max. effluent H₂O concentration 14 ppb
    Refillable moisture trap, 400 ml, removal capacity 72 g, max. effluent H₂O concentration 14 ppb
    HEWLMT400-2
    MT400-2
    05
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 286,00 €
    286,00 €
    New Customer Promotional Price
    1 * 1 items
       286,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Fitting size
    1/4"