Dithiothreitol (DTT, Cleland's reagent) 2 mol/l aqueous solution
Catalog # 786-227
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:5
- Seq:0005
- Environmentally Preferable:
Specifications
About this item
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
:
:154.25 g/mol
EINECS:222-468-7
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877