VWR®, Bovine serum albumin (BSA)
Supplier: VWR
VWR®
0903-10GEA 339 EUR
0903-10G
VWR®, Bovine serum albumin (BSA)
Proteins and Peptides
Cold alcohol isolation suitable for sensitive biochemical and diagnostic assays. DNase and RNase free, virtually free of globulins and other interfering contaminants.
Crystalline.This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays.