Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "in-vivo-imaging-dyes"
https://si.vwr.com/stibo/smallweb/std.lang.all/53/18/18945318.jpg
SearchPresentationType-HORIZONTAL
SearchResultCount:"8"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Biotium
Description:   CF™ 750 free acid
Supplier:  Biotium
Description:   D-Luciferin is a substrate for the enzyme firefly luciferase. Chemiluminescence at 560 nm is generated when luciferin is oxidised by an ATP-dependent ...
Supplier:  Biotium
Description:   Near-infrared (near-IR) dyes offer important advantages over traditional visible light dyes. Because cellular or tissue components produce minimal aut...
Supplier:  Biotium
Description:   VivoBrite™ Antibody Labelling kits include amine-reactive near-IR CF™ dye and all other components required for antibody labeling and purification.
Supplier:  Biotium
Description:   Chromogenic and fluorogenic peroxidase substrates.
Supplier:  Biotium
Description:   β-Glucuronidase substrates are easier to detect against the background of plant cells.
Supplier:  Biotium
Description:   Chromogenic and fluorogenic beta-galactosidase substrates.
Catalog Number: (10026.)
Supplier:  Biotium
Description:   X-β-D-cellobioside (5-Bromo-4-chloro-3-indolyl-beta-D-cellobioside) is a chromogenic substrate for β-cellobiosidase. Enzymatic cleavage of the substra...
UOM:  1 * 25 mg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 8