Order Entry
Safety cabinets for flammable, BSC classic
Safety cabinets for flammable, BSC classic
Certificates
Safety cabinets for flammable, BSC classic
Product Information
  • Catalog #
    Supplier #
    Seq
    Doors
    W×D×H
    W×D×H int.
    Weight
    Pk
    Availability
    Price
  • Fire resistant cabinet BSC 2-CL, storage capacity: 400 l (door dimension 1250×2015 mm)
    Fire resistant cabinet BSC 2-CL, storage capacity: 400 l (door dimension 1250×2015 mm)
    LACOC64-2000-B
    C64-2000-B
    01
    1
    2170×2350×2510 mm
    1760×2100×2070 mm
    1500 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 3-CL, storage capacity: 660 l (door dimension 1250×2015 mm)
    Fire resistant cabinet BSC 3-CL, storage capacity: 660 l (door dimension 1250×2015 mm)
    LACOC64-2001-B
    C64-2001-B
    02
    1
    3000×2350×2510 mm
    2760×2100×2070 mm
    2100 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 3-CL, storage capacity: 660 l (door dimension 1800×2015 mm)
    Fire resistant cabinet BSC 3-CL, storage capacity: 660 l (door dimension 1800×2015 mm)
    LACOC64-2031-B
    C64-2031-B
    03
    2
    3000×2350×2510 mm
    2760×2100×2070 mm
    2100 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 4-CL, storage capacity: 1000 l (door dimension 1250×2015 mm)
    Fire resistant cabinet BSC 4-CL, storage capacity: 1000 l (door dimension 1250×2015 mm)
    LACOC64-2002-B
    C64-2002-B
    04
    1
    4000×2350×2510 mm
    3760×2100×2070 mm
    2550 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 4-CL, storage capacity: 1000 l (door dimension 1800×2015 mm)
    Fire resistant cabinet BSC 4-CL, storage capacity: 1000 l (door dimension 1800×2015 mm)
    LACOC64-2032-B
    C64-2032-B
    05
    2
    4000×2350×2510 mm
    3760×2100×2070 mm
    2550 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 5-CL, storage capacity: 1100 l (door dimension 1250×2015 mm)
    Fire resistant cabinet BSC 5-CL, storage capacity: 1100 l (door dimension 1250×2015 mm)
    LACOC64-2003-B
    C64-2003-B
    06
    1
    5000×2350×2510 mm
    4760×2100×2070 mm
    2950 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 5-CL, storage capacity: 1100 l (door dimension 1800×2015 mm)
    Fire resistant cabinet BSC 5-CL, storage capacity: 1100 l (door dimension 1800×2015 mm)
    LACOC64-2033-B
    C64-2033-B
    07
    2
    5000×2350×2510 mm
    4760×2100×2070 mm
    2950 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 6-CL, storage capacity: 1300 l (door dimension 1250×2015 mm)
    Fire resistant cabinet BSC 6-CL, storage capacity: 1300 l (door dimension 1250×2015 mm)
    LACOC64-2004-B
    C64-2004-B
    08
    1
    6000×2350×2510 mm
    5760×2100×2070 mm
    3400 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 6-CL, storage capacity: 1300 l (door dimension 1800×2015 mm)
    Fire resistant cabinet BSC 6-CL, storage capacity: 1300 l (door dimension 1800×2015 mm)
    LACOC64-2034-B
    C64-2034-B
    09
    2
    6000×2350×2510 mm
    5760×2100×2070 mm
    3400 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 7-CL, storage capacity: 1600 l (door dimension 1800×2015 mm)
    Fire resistant cabinet BSC 7-CL, storage capacity: 1600 l (door dimension 1800×2015 mm)
    LACOC64-2035-B
    C64-2035-B
    10
    2
    7000×2350×2510 mm
    6760×2100×2070 mm
    3900 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Fire resistant cabinet BSC 8-CL, storage capacity: 1800 l (door dimension 1800×2015 mm)
    Fire resistant cabinet BSC 8-CL, storage capacity: 1800 l (door dimension 1800×2015 mm)
    LACOC64-2036-B
    C64-2036-B
    11
    2
    8000×2350×2510 mm
    7760×2100×2070 mm
    4400 kg
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Accessories