VWR® LB (Lennox), Agar, Tissue Culture Grade
Supplier: Avantor
VWR®
K497-10PKEA 152 EUR
K497-10PK
VWR® LB (Lennox), Agar, Tissue Culture Grade
Media for Microbiology Dehydrated Media for Microbiology
Used for the propagation and maintenance of E. coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
  • Provides a solid growth medium
  • Used in molecular genetic studies
  • Used as a solidification agent
K497-10PK contains 10 individual pouches. Dissolve the contents of a single pouch in deionised water and autoclave to prepare one L of LB agar (Lennox formulation).