UHPLC columns, Pinnacle® DB Cyano (USP L10)
Supplier: Restek
Certificates
About this item
Highly base-deactivated spherical silica manufactured by Restek. Cyano bonding. Suitable for analyses of a wide range of compounds, from acidic through slightly basic. Also useful for confirmation of analyses on a C18 or C8 column. Can be used in normal phase or reversed phase mode of separation.
Specifications
- Catalog No:
- REST9416512
- REST9416511
- REST9416510
- 554-1563
- REST9416579
- REST9416578
- 554-1562
- REST9416577
- 554-1561
- REST9416575
- REST9416573
- REST9416572
- REST9416513
- REST9416571
- REST9416570
- REST9416569
- REST9416568
- REST9416567
- REST9416565
- REST9416563
- REST9416562
- REST9416561
- REST9416560
- REST9416573-700
- REST9416575-700
- REST9416562-700
- REST9416563-700
- REST9416565-700
- REST9416572-700
- REST941650222
- REST9416559
- REST941650220
- REST9416558
- REST941650212
- REST9416557
- REST941650210
- REST9416555
- REST9416553
- REST9416552
- REST9416551
- REST9416550
- REST9416535
- REST9416533
- REST9416552-700
- REST9416553-700
- REST9416555-700
- REST9416525-700
- REST9416532-700
- REST9416533-700
- REST9416535-700
- REST9416515-700
- REST9416522-700
- REST9416523-700
- REST941655E
- REST941656E
- REST941657E
- REST9416532
- REST9416531
- REST9416525
- REST9416523
- REST9416522
- REST9416521
- REST9416519
- REST9416518
- REST9416517
- REST9416515
- REST9416512-700
- REST9416513-700
- REST941651E
- REST941652E
- REST941653E
- Carbon load:
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
- L10, L10
Specifications
With trident integral inlet fittings
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- REST9416562-7009416562-700AnalyticalCyano140 Å5 µm2,10 mm150 mmLog in for availability1 * 1 items643,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416572-7009416572-700AnalyticalCyano140 Å5 µm2,10 mm250 mmLog in for availability1 * 1 items686,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416563-7009416563-700AnalyticalCyano140 Å5 µm3,20 mm150 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416573-7009416573-700AnalyticalCyano140 Å5 µm3,20 mm250 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416565-7009416565-700AnalyticalCyano140 Å5 µm4,60 mm150 mmLog in for availability1 * 1 items810,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416575-7009416575-700AnalyticalCyano140 Å5 µm4,60 mm250 mmLog in for availability1 * 1 items1 064,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416512-7009416512-700AnalyticalCyano140 Å5 µm2,10 mm100 mmLog in for availability1 * 1 items603,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416522-7009416522-700AnalyticalCyano140 Å5 µm2,10 mm200 mmLog in for availability1 * 1 items654,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416532-7009416532-700AnalyticalCyano140 Å5 µm2,10 mm30 mmLog in for availability1 * 1 items501,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416552-7009416552-700AnalyticalCyano140 Å5 µm2,10 mm50 mmLog in for availability1 * 1 items563,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416513-7009416513-700AnalyticalCyano140 Å5 µm3,20 mm100 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416523-7009416523-700AnalyticalCyano140 Å5 µm3,20 mm200 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416533-7009416533-700AnalyticalCyano140 Å5 µm3,20 mm30 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416553-7009416553-700AnalyticalCyano140 Å5 µm3,20 mm50 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416515-7009416515-700AnalyticalCyano140 Å5 µm4,60 mm100 mmLog in for availability1 * 1 items672,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416525-7009416525-700AnalyticalCyano140 Å5 µm4,60 mm200 mmLog in for availability1 * 1 items889,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416535-7009416535-700AnalyticalCyano140 Å5 µm4,60 mm30 mmLog in for availability1 * 1 items501,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10 - REST9416555-7009416555-700AnalyticalCyano140 Å5 µm4,60 mm50 mmLog in for availability1 * 1 items568,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 7,5Temperature limit80 °CUSP codeL10, L10
Without trident integral inlet fittings
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165129416512AnalyticalCyano140 Å5 µm2,10 mm100 mmLog in for availability1 * 1 items587,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165629416562AnalyticalCyano140 Å5 µm2,10 mm150 mmLog in for availability1 * 1 items632,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165729416572AnalyticalCyano140 Å5 µm2,10 mm250 mmLog in for availability1 * 1 items677,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165659416565AnalyticalCyano140 Å5 µm4,60 mm150 mmLog in for availability1 * 1 items760,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165759416575AnalyticalCyano140 Å5 µm4,60 mm250 mmLog in for availability1 * 1 items969,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165579416557Semi-preparativeCyano140 Å5 µm10,00 mm50 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165679416567Semi-preparativeCyano140 Å5 µm10,00 mm150 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165779416577Semi-preparativeCyano140 Å5 µm10,00 mm250 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165589416558Semi-preparativeCyano140 Å5 µm21,20 mm50 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165689416568Semi-preparativeCyano140 Å5 µm21,20 mm150 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165789416578Semi-preparativeCyano140 Å5 µm21,20 mm250 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165699416569PreparativeCyano140 Å5 µm30,00 mm150 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165799416579PreparativeCyano140 Å5 µm30,00 mm250 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165109416510PreparativeCyano140 Å5 µm50,00 mm100 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165609416560PreparativeCyano140 Å5 µm50,00 mm150 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165709416570PreparativeCyano140 Å5 µm50,00 mm250 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165599416559PreparativeCyano140 Å5 µm30,00 mm50 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165529416552AnalyticalCyano140 Å5 µm2,10 mm50 mmLog in for availability1 * 1 items546,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165359416535AnalyticalCyano140 Å5 µm4,60 mm30 mmLog in for availability1 * 1 items489,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165559416555AnalyticalCyano140 Å5 µm4,60 mm50 mmLog in for availability1 * 1 items546,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165509416550PreparativeCyano140 Å5 µm50,00 mm50 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165329416532AnalyticalCyano140 Å5 µm2,10 mm30 mmLog in for availability1 * 1 items489,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165229416522AnalyticalCyano140 Å5 µm2,10 mm200 mmLog in for availability1 * 1 items642,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941653E941653EAnalyticalCyano140 Å5 µm3,00 mm30 mmLog in for availability1 * 1 items489,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941655E941655EAnalyticalCyano140 Å5 µm3,00 mm50 mmLog in for availability1 * 1 items546,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941651E941651EAnalyticalCyano140 Å5 µm3,00 mm100 mmLog in for availability1 * 1 items587,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941656E941656EAnalyticalCyano140 Å5 µm3,00 mm150 mmLog in for availability1 * 1 items644,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941652E941652EAnalyticalCyano140 Å5 µm3,00 mm200 mmLog in for availability1 * 1 items748,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941657E941657EAnalyticalCyano140 Å5 µm3,00 mm250 mmLog in for availability1 * 1 items756,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165159416515AnalyticalCyano140 Å5 µm4,60 mm100 mmLog in for availability1 * 1 items587,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165259416525AnalyticalCyano140 Å5 µm4,60 mm200 mmLog in for availability1 * 1 items833,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165179416517Semi-preparativeCyano140 Å5 µm10,00 mm100 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165189416518Semi-preparativeCyano140 Å5 µm21,20 mm100 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST94165199416519PreparativeCyano140 Å5 µm30,00 mm100 mmLog in for availability1 * 1 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10
Guard cartridges
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- REST941650222941650222Guard cartridgeCyano140 Å5 µm2,10 mm20 mmLog in for availability1 * 2 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941650220941650220Guard cartridgeCyano140 Å5 µm4,00 mm20 mmLog in for availability1 * 2 items0,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941650212941650212Guard cartridgeCyano140 Å5 µm2,10 mm10 mmLog in for availability1 * 3 items277,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10 - REST941650210941650210Guard cartridgeCyano140 Å5 µm4,00 mm10 mmLog in for availability1 * 3 items277,00 €Specifications:
More
Specifications
Environmentally PreferableCarbon load4%EndcappedYespH range2,5 - 8Temperature limit80 °CUSP codeL10, L10