Order Entry
Rotors for MIKRO 220 centrifuge
Rotors for MIKRO 220 centrifuge
Supplier:  Hettich
Certificates
undefined
Rotors for MIKRO 220 centrifuge
Supplier:  Hettich
Product Information
  • Catalog #
    Supplier #
    Seq
    Pk
    Availability
    Price
  • Fixed angle rotor 1015, 12×15 ml conical tubes, 6000 min-1, 4146 ×g, for use with MIKRO 220 / 220R
    Fixed angle rotor 1015, 12×15 ml conical tubes, 6000 min-1, 4146 ×g, for use with MIKRO 220 / 220R
    HETT1015
    1015
    01
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 425,00 €
    425,00 €
    New Customer Promotional Price
    1 * 1 items
       425,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Fixed angle rotor 2218-A, 12×0,2/2 ml conical tubes, 18000 min-1, 25718 ×g, for use with MIKRO 220 / 220R
    Fixed angle rotor 2218-A, 12×0,2/2 ml conical tubes, 18000 min-1, 25718 ×g, for use with MIKRO 220 / 220R
    HETT2218-A
    2218-A
    02
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • Fixed angle rotor, 6×50 ml conical tubes, 6000 min⁻¹, 4025×g, for use with MIKRO 220 R
    Fixed angle rotor, 6×50 ml conical tubes, 6000 min⁻¹, 4025×g, for use with MIKRO 220 R
    521-1896
    1016
    03
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 639,00 €
    639,00 €
    New Customer Promotional Price
    1 * 1 items
       639,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Fixed angle rotor 1158-L, 48×1,5/2,0 ml (2 rows), 14000 min⁻¹, 21255×g, with biocontainment lid
    Fixed angle rotor 1158-L, 48×1,5/2,0 ml (2 rows), 14000 min⁻¹, 21255×g, with biocontainment lid
    521-1186
    1158-L
    04
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 886,00 €
    886,00 €
    New Customer Promotional Price
    1 * 1 items
       886,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Fixed angle rotor 1160, 6-place, for PCR strips, 14000 min⁻¹, 18845×g
    Fixed angle rotor 1160, 6-place, for PCR strips, 14000 min⁻¹, 18845×g
    521-1522
    1160
    05
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • Fixed angle rotor 1189-A, 30×1,5/2,0 ml, 14000 min⁻¹, 21255×g, with biocontainment lid
    Fixed angle rotor 1189-A, 30×1,5/2,0 ml, 14000 min⁻¹, 21255×g, with biocontainment lid
    521-1187
    1189-A
    06
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 620,00 €
    620,00 €
    New Customer Promotional Price
    1 * 1 items
       620,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Fixed angle rotor 1195-A, 24×1,5/2,0 ml, 18000 min⁻¹, 31514×g, with biocontainment lid
    Fixed angle rotor 1195-A, 24×1,5/2,0 ml, 18000 min⁻¹, 31514×g, with biocontainment lid
    521-1188
    1195-A
    07
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 496,00 €
    496,00 €
    New Customer Promotional Price
    1 * 1 items
       496,00 €
    Specifications:
    Specifications
    Environmentally Preferable