Centrifuge, refrigerated, floor-standing, Sorvall™ RC3BP™ Plus
Supplier: Thermo Fisher Scientific
Sorvall™
521-1983EA 37280 EUR
521-1983 521-1984 521-1985
Centrifuge, refrigerated, floor-standing, Sorvall™ RC3BP™ Plus
Centrifuges Floor-Standing Centrifuges
This low-speed centrifuge combines outstanding convenience with exceptional ease of use to expedite daily processing.
  • Low deck height, quiet operation, minimal heat output and increased refrigeration efficiency
  • Automatic lid-latch system protects users and enables effortless opening and closing of the centrifuge
  • Slow start/stop mode: Users can select from 11 acceleration and deceleration profiles
  • Easy-to-use operator interface makes set-up and operation straightforward, even for novice users
  • 15-program memory provides one-touch recall of commonly used protocols
  • Security key prevents unauthorised access
Suitable for a wide range of applications including blood banking, low-speed separations and general, large-batch and high-volume processing. The available rotors, adapters and carriers provide versatility for diverse separation applications.
Informace pro objednávání: Supplied without rotors. Rotors and accessories can be ordered separately.