This low-speed centrifuge combines outstanding convenience with exceptional ease of use to expedite daily processing.
- Low deck height, quiet operation, minimal heat output and increased refrigeration efficiency
- Automatic lid-latch system protects users and enables effortless opening and closing of the centrifuge
- Slow start/stop mode: Users can select from 11 acceleration and deceleration profiles
- Easy-to-use operator interface makes set-up and operation straightforward, even for novice users
- 15-program memory provides one-touch recall of commonly used protocols
- Security key prevents unauthorised access
Suitable for a wide range of applications including blood banking, low-speed separations and general, large-batch and high-volume processing. The available rotors, adapters and carriers provide versatility for diverse separation applications.
Informace pro objednávání: Supplied without rotors. Rotors and accessories can be ordered separately.